Sterilisation indicating labels for thermal transfer, series R4300
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
149273 149272
BRDY149273EA 202.56 EUR
BRDY149273 BRDY149272
Sterilisation indicating labels for thermal transfer, series R4300
Etiketten
This B-7425-AC material is a steam sterilisation indicating polypropylene label with a preprinted header that displays 'STERILIZED' when exposed to autoclave conditions. It's ideal for use in sample tracking, inventory management, general tracking, and identification of laboratory materials that require sterilisation.
  • Die-cut labels are on a roll
  • Application: general identification, laboratory labelling, temperature indicating labelling
  • This R4300 label has a matt finish
  • Service temperature: –80 to 70 °C
  • Thickness: 0,102 mm
  • Autoclavable and cryogenic storage are the material's properties
This node contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates. This node contains only supplies sold with the ribbon seperate, it is not in the same enclosure as the tape/sleeves.
This economical solution for identifying vials, tubes, racks and flasks adheres to stainless steel, glass and polypropylene, and can withstand temperatures from –196 to 121°C.