Label printers with peel function and vial label applicator, industrial thermal transfer benchtop, BradyPrinter i7100
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
307652 307658 307653 307659
BRDY307652EA 5082 EUR
BRDY307652 116-0938 BRDY307653 BRDY307659
Label printers with peel function and vial label applicator, industrial thermal transfer benchtop, BradyPrinter i7100
Printers Label Printers
The BradyPrinter i7100 industrial thermal transfer benchtop printer is ideal for customers requiring reliable high volume printing on lots of different materials within the aerospace, defence and mass transit, electronics, automotive, logistics, electrical, voice-datacom and LAB markets.
  • Prints labels and then applies them to wires or cables
  • Two step process
  • Laboratory labelling application
  • Lithium-manganese battery
  • Colour: Monocolour
It guarantees accurate printing on sleeves, tags, labels, tapes, cable markers and much more. This printer comes with either a Brady WorkStation Basic Design Software Suite or Laboratory ID Suite, enabling you to immediately start designing professional labels.
Bar-code: code 39; code 93; code 39 Full ASCII; code 128 A; code 128 B; code 128 C; EAN 8; EAN13; EAN/UCC 128/GS1-128; EAN/UPC; FIM; HIBC; interleaved 2/5; ident and lead code of Deutsche Post AG; Codabar; JAN8; JAN 13; MSI; Plessey; Postnet; RSS14; UPC A; UPC E; UPC E0; Aztec; Codablock F; DataMatrix; PDF417; Micro PDF417; UPS MaxiCode; QR code; RSS 14 truncated, limited stacked, stacked and stacked omnidirectional; EAN/GS1 DataMatrix; GS1 DataBar.
Languages: Model 307653 only: Arabic Dutch; Bulgarian; Chinese (Simplified); Croatian; Czech; Danish; Dutch; English (US); Farsi; Finnish; French; German; Greek; Hungarian; Italian; Korean; Lithuanian; Macedonian; Norwegian (Bokmål); Polish; Portuguese; Romanian; Russian; Serbian; Slovenian; Spanish (Mexico); Spanish; Swedish; Thai; Turkish; Others: Arabic (Egypt); Bulgarian; Croatian; Czech; Danish; Dutch; English; Finnish; French; German; Greek; Hungarian; Italian; Lithuanian; Macedonian; Norwegian (Bokmål); Persian (Farsi); Polish; Portuguese; Romanian; Russian; Serbian; Slovenian; Spanish; Swedish; Turkish.
Leveringsinformatie: The i7100 printer comes in a cardboard box with the Peel and Present module pre-installed. It ships with a vial label applicator, an EU or UK power cord, a Brady Workstation Basic Suite or Brady Workstation Laboratory ID Suite, two empty ribbon cores, a USB cable, a product CD holding the driver, the user manual in languages, the configuration/settings manual in English and a printed user manual in English.