Ionenuitwisselingschromatografiehars, J.T. Baker® BAKERBOND® PolyQUAT multimode
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7603-01EA 208.21 EUR
7603-01 7603-02 7603-03 7603-04 7603-05
Ionenuitwisselingschromatografiehars, J.T. Baker® BAKERBOND® PolyQUAT multimode
Sorbenten voor chromatografie Sorbenten voor HPLC
The J.T.Baker® BAKERBOND® PolyQUAT multimode ion exchange (IEX) chromatography resin functions as a strong anion exchange resin with improved selectivity for superior separation in process chromatography applications.
Powered by a proprietary surface chemistry combining strong anion exchange functionality with weak anionic exchange sites and hydrophilicity, BAKERBOND® PolyQUAT multimode cation exchange chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Ontworpen en vervaardigd door Avantor volgens de hoge normen die zijn vastgesteld voor onze J.T.Baker® merkchemicaliën, kan de BAKERBOND® PolyQUAT ionenuitwisselingshars worden gebruikt met andere J.T.Baker® BAKERBOND® harsen om het scheidend vermogen van uw chromatografische processtappen te.
Bestelinformatie: 1 ml and 5 ml lab columns are also available under complementary products.
Order Now
Specificaties
Environmentally Preferable
Werk-pH 2-14