Urinal mats with adhesive backing, PIG®
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GRP7001-BK GRP7601-BK GRP7002-BK GRP7602-BK
115-2580EA 57.5 EUR
115-2580 115-2581 115-2582 115-2583
Urinal mats with adhesive backing, PIG®
Vloermatten Kleefmatten
Keep your restrooms cleaner and provide better drip and splash protection with these adhesive-back urinal mats.
  • Sink and dryer mats capture water to keep floors dry in front of sinks and under hand dryers and towel dispensers
  • Help control slip hazards around drinking fountains, showers, and other wet trouble spots
  • Permanently infused antimicrobial technology prevents the growth of odor-causing bacteria and mold on the mats to keep restrooms cleaner and fresher smelling
  • Proprietary, full-coverage adhesive backing keeps mats in place underfoot with no shifting, curling or flipping up to stop slip and trip hazards; peels up easily
  • Mats stick to your floor and won't budge while you clean them; simply mop them with the rest of the floor
  • Durable, Hi-Trac™ surface stands up to daily cleaning and foot traffic
  • Absorbs water and urine
EFM™ Absorbent technology captures water on contact, wicks it away and dries out quickly. Impermeable backing layers stop liquid pass-through to prevent stains and grout damage.
Release paper on the bottom of the mat should be removed prior to installation.
Certificaten: Tested and certified by the National Floor Safety Institute (NFSI) as a high-traction surface which can reduce slip and fall claims by as much as 90% when used as part of a floor safety program.
Opgelet: Mats take on the properties of absorbed liquids - always take proper precautions when handling saturated absorbents.