Mat rolls in display box, PIG®
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAT1991-EN MAT1991-FR MAT1991-IT MAT1991-DE
NEWPMAT1991-ENEA 448 EUR
NEWPMAT1991-EN NEWPMAT1991-FR NEWPMAT1991-IT NEWPMAT1991-DE
Mat rolls in display box, PIG®
Absorptiematerialen en bescherming tegen morsen
This handy display box holds 12 conveniently-sized mat rolls, perfect for garages, workshop floors, work benches, tool boxes, machinery, vehicles and service bays.
  • Eight layers of 100% polypropylene are thermally bonded to make PIG® Mat strong; won't rip, tear or fray even when saturated
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave behind liquids or fibre residue
  • Light-weight construction is ideal for small tasks and clean-ups
  • Perforated every 25 cm, so you only use what you need
Perfect for garages, workshop floors, work benches, tool boxes, machinery, vehicles and service bays. Roll size makes it easy to store and carry. Easy-tear perforations let you take only what you need.
Absorbs oils, grease, water, diesel and fuels, solvents, coolants, paints and inks. Superior durability, wicking and retention. Made from up to 25% recycled content.