HPLC supply-sets, V2.0
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
307300 307301 307303 307304 307305 307306 307312 307313 307314 307347
SCTV307300EA 108 EUR
SCTV307300 SCTV307301 SCTV307303 SCTV307304 SCTV307305 SCTV307306 SCTV307312 SCTV307313 SCTV307314 SCTV307347
HPLC supply-sets, V2.0
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
The HPLC Supply Sets, V2.0 of the premium manufacturer SCAT are attractively priced combinations of equipment for contamination-free supply of HPLC systems with solvents. The sets each consist of a laboratory bottle with GL 45 thread, a SCAT SafetyCap as well as matching capillaries, suction filters and air valves.
  • Customised system with high quality set components
  • SafetyCaps of the premium manufacturer SCAT
  • Laboratory bottles of DURAN® brand
  • Price attractive in comparison with single purchase
  • Ready for use, no further components necessary
SCAT offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other. This makes SCAT the No. 1 system for solvent safety!
Bestelinformatie: HPLC Supply-Set I, V2.0: 1×1,5 m capillary 3,2 mm Ext. Ø., 1×PTFE suction filter + air valve
HPLC Supply-Set II, V2.0: 2×1,5 m capillaries 3,2 mm Ext. Ø, 2×PTFE suction filters + air valve
HPLC Supply-Set III, V2.0: 3×1,5 m capillaries 3,2 mm Ext. Ø, 3×PTFE suction filters + air valve
All components can be reordered individually.