Ladles, long handle, disposable, LaboPlast®/SteriPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5378-5022 5378-5021
331-0358EA 24.6 EUR
331-0358 331-0359
Ladles, long handle, disposable, LaboPlast®/SteriPlast®
Scheppen
The ladle is suitable for taking samples of liquids and viscous media as well as powders and granulates.
  • PS, white
  • Individually packaged and optionally sterilised
  • Reaches into any corner
  • Handle length 300 mm
  • For single use
  • Clean room manufactured (clean room class 7)
The ladle has a very smooth bottom and rounded corners. This makes it possible to reach into any corner of a container or to take medium directly from the bottom of the container. A long handle with indentations for fingers ensures a firm hold and precise work. The disposable ladle can be used in the pharmaceutical industry, chemistry and cosmetics and the food and animal feed industries.
Certificaten: Complies with EU food requirements and FDA regulations.