Serial extraction unit, R 306/R 306 S, behrotest®
Leverancier: BEHR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B00218434 B00218444
BEHRB00218434EA 5300 EUR
BEHRB00218434 BEHRB00218444
Serial extraction unit, R 306/R 306 S, behrotest®
Extractors Soxhlet-extractors en Soxhlet-type extractors
Complete compact extraction system with basic rack, heating device, holder, hoses and glassware (reaction flasks, extractor, Dimroth condenser for extraction).
  • Practical holders for condenser and extraction adapter for secure placement between extractions
  • Extractor sizes from 30 to 1000 ml
  • Condenser with screw connections
Infinitely variable heating regulation. After the extraction cycle, the compact system removes the solvent, by means of a discharge valve on the extractor directly into the reservoir. Behr equipment for Soxhlet extraction complies with the various requirements of daily laboratory routines.
With behr hydrolysis units, acid digestion is possible before extraction (determination of total fat content according to Weibull and Stoldt).
Weight (in kg): 23
Sample positions: 6
Flask size (in ml): 100
Extractor size (in ml): 30
Leveringsinformatie: Supplied with sample package, extraction sleeves and boiling stones.