Sampling scoop, long handle, disposable Bio
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5379-1024 5379-0024 5379-1025 5379-0025
331-0354EA 34.2 EUR
331-0354 331-0355 331-0356 331-0357
Sampling scoop, long handle, disposable Bio
Scheppen
The sampling scoop can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even with little content or to reach through narrow openings. With the straight bottom of the scoop, the medium can be scooped up directly from the bottom of the container.
  • Product made from renewable raw materials
  • Bio-PE, white
  • Individually packaged and optionally sterilised
  • Handle length 300 mm
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage,Duurzame materialen
Milieu certificering ACT-Label