Ladle, long handle, disposable Bio
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5379-1026 5379-0026
331-0362EA 28.6 EUR
331-0362 331-0363
Ladle, long handle, disposable Bio
Scheppen
The ladle is suitable for taking samples of liquids and viscous media as well as powders and granulates. The ladle has a very smooth bottom and rounded corners. This makes it possible to reach into any corner of a container or to take medium directly from the bottom of the container. A long handle with indentations for fingers ensures a firm hold and precise work.
  • Product made from renewable raw materials
  • Bio-PE, white
  • Individually packaged and optionally sterilised, for single use
  • Reaches into any corner
  • Handle length 300 mm
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
The disposable ladle can be used in the pharmaceutical industry, chemistry and cosmetics and the food and animal feed industries.