Sampling palette knife spatula, disposable Bio, LaboPlast®Bio/SteriPlast®Bio
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5379-1035 5379-0035
331-0372EA 75.3 EUR
331-0372 331-0373
Sampling palette knife spatula, disposable Bio, LaboPlast®Bio/SteriPlast®Bio
Spatels
The sampling palette knife spatula can be used to apply, distribute or evenly spread viscous media such as pastes and creams. The palette knife spatula reaches easily into the corners and curves of various containers and is suitable for scraping out bowls, removing residue or wiping off Bürkle’s volumetric and dosing spoons.
  • Product made from renewable raw materials
  • Bio PE, white, flexible and break-proof
  • L×W 192×20 mm
  • Optionally packaged individually and sterilised
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA