Chemical resistant gloves, nitrile, KleenGuard® G80
Leverancier: KIMBERLY CLARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25622 25623 25624 25625
KIMB25622EA 104 EUR
KIMB25622 KIMB25623 KIMB25624 KIMB25625
Chemical resistant gloves, nitrile, KleenGuard® G80
Handschoenen
These gloves are suitable for chemical handling, manufacturing, construction and agriculture, contract cleaning and janitorial and public service.
  • Unlined, reducing the risk of lint contamination
  • Extended length to protect the wrist and forearm
  • Sand patch structure on fingers and palm for improved wet and dry grip
Gloves provide protection against oils, greases, acids, caustics and solvents.
Certificaten: Cat. III; AQL 0,65