Wipers, WYPALL® X80
Leverancier: KIMBERLY CLARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8294 8295
KIMB8294EA 93.7 EUR
KIMB8294 KIMB8295
Wipers, WYPALL® X80
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Doekjes voor algemeen gebruik
The WypAll® X80 Range has been specially designed for heavy-duty, rough surface wiping, where strength and bulk are key requirements. WypAll® X80 cloths do not tear and maintaining consistency in quality and size.
  • Cloths are strong and tear-proof, maintaining integrity and size
  • Perfect for use in industrial manufacturing
  • Ideal for portability or small work spaces
  • Hydroknit technology ensure each cloth is strong, durable and tear-proof
The WypAll® range is designed to help improve productivity and efficiency whilst reducing waste.
Verpakking: More Product Information