Wipe rolls, WypAll® L10 centrefeed food and hygiene
Leverancier: KIMBERLY CLARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7255 7256
KIMB7255EA 91.2 EUR
KIMB7255 KIMB7256
Wipe rolls, WypAll® L10 centrefeed food and hygiene
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Doekjes voor algemeen gebruik
WypAll® L10 professional wiping paper products are designed for the specific environments where they will be used, helping enable you to deliver on your wiping task every time.
  • Single use wiping paper
  • Suitable for food contact
  • Centrefeed paper rolls are designed for use with centrefeed dispensers
  • WypAll® wiping paper is designed to be used once with detergent or disinfectant
Dry cleaning wipes designed for light duty hygiene wiping and perfect for tasks like cleaning food preparation and service surfaces. Embossing technology in the wiping paper gives these cleaning wipes a great balance of performance and value.
These dry cleaning wipes have been tested as compatible for use in spray and wipe application with peracetic acid, hydrogen peroxide, and quats (quaternary ammonium compounds). Spray and wiping compatibility helps to ensure workers can wipe own work spaces, vehicles, desks, seating, and food prep areas before and after use. Spray and wipe also helps to enable production workers to clean down/sanitise individual workstations before hand-off to next shift. For environments where fully saturated wiping is a requirement, these WypAll® rolls are tested as compatible with soap/detergent.