Labelling tapes, thermal transfer printable tapes
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
150608 150609 150610 150611 150612 150613 150614 150615
BRDY150608EA 262.86 EUR
BRDY150608 BRDY150609 BRDY150610 BRDY150611 BRDY150612 BRDY150613 BRDY150614 BRDY150615
Labelling tapes, thermal transfer printable tapes
Etiketten
Continuous tapes for printers.
  • Choice of repositionable rubber or permanent acrylic adhesives
  • Continuous tape is not cut to length, it gets cut by the printer
  • This node contains only supplies sold with the ribbon seperate, it is not in the same enclosure as the tape/sleeves
Vinyl labelling tapes feature a repositionable, rubber based adhesive and semi-gloss finish. Vinyl tapes are ideal for general identification, laboratory labelling, and wire and cable labelling applications. These tapes should be used with IP-R6206 and R6206 ribbons, and areMore Product Information