VWR®, Fast extract DNA solution
Leverancier: VWR ChemicalsTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
The fast extract DNA solution provides rapid and efficient extraction of PCR-ready DNA from mammalian and plant tissues. DNA is ready in eight minutes. The one-reagent protocol is divided into two simple heating steps, which can be directly followed by PCR analysis, such as screening and genotyping.
- One-reagent set-up
- Rapid eight-minute protocol
- PCR-ready DNA
- DNA extracts from mammalian tissues, plant materials, and bacteria
- Non-toxic reagents
- Automation-friendly
DNA can be extracted from many different sample types e.g. mouse tails or ears, saliva, bacteria, plant material, and mammalian tissues . The one-reagent DNA extraction set-up is easily scaled and can be conducted by robotic automation platforms. Depending on the sample size, the DNA extraction can be performed in PCR tubes or in 1,5 ml tubes, using either a thermocycler or a heating block.
Prep size: 100 µl
Method/format: DNA extraction