Magnesium(II) gluconate hydrate
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: D-Gluconic acid hemimagnesium salt hydrate, Magnesium D-gluconate hydrate, Gluconic acid magnesium(II) salt hydrate, D-Gluconic acid magnesium salt hydrate
464830250 464831000
ACRO464830250EA 33.2 EUR
ACRO464830250 ACRO464831000
Magnesium(II) gluconate hydrate
Magnesium(II) gluconate hydrate
  • Bioactive
Formule: C₆H₁₂O₇·xH₂O
MW: 414.60 g/mol (anhy)
Smeltpunt: 169.7…171.9 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150971
CAS nummer: 3632-91-5
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration Complexometric 98.0 to 102.0 % (on dry substance)
Water ≤13.0 %