Clemastine fumarate
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: (R)-2-(2-((R)-1-(4-Chlorophenyl)-1-phenylethoxy)ethyl)-1-methylpyrrolidine fumarate
465420010 465422500
ACRO465420010EA 615 EUR
ACRO465420010 ACRO465422500
Clemastine fumarate
Clemastine fumarate
  • Bioactive
Formule: C₂₅H₃₀ClNO₅
MW: 459.9600 g/mol
Smeltpunt: 158…162 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00137486
CAS nummer: 14976-57-9
Order Now
Specification Test Results
Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %
Specific optical rotation +15° to +18° (20°C, 589 nm) (10 mg/ml methanol)