Indacaterol maleate

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

464650010 464652500
ACRO464650010EA 446 EUR
ACRO464650010 ACRO464652500
Indacaterol maleate
Indacaterol maleate

  • Bioactive

Formule: C₂₄H₂₈N₂O₃·C₄H₄O₄
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD20526769
CAS nummer: 753498-25-8

Order Now

Specification Test Results

Appearance Powder
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR