Lead-free steam indicator tapes, Comply™
Leverancier: 3M MEDICAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M1322-24 M1322-18 1322-12
489-0107EA 22.4 EUR
489-0107 817-0118 MMME1322-12
Lead-free steam indicator tapes, Comply™
Sterilisatie-indicatoren
An exposure monitor that securely seals packs, and provides a visual assurance that the package has been exposed to the steam sterilisation process.
  • Not made with natural rubber latex
  • Trusted, high-quality adhesive
  • Lead-free ink means no hazardous waste issues
  • Stretchable backing that minimises tape peel off during sterilisation
  • Can be written on or labeled with pre-printed labels
As an exposure monitor, Comply™ Indicator tapes securely seal packs and enable CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. The chemical indicator ink, printed in a diagonal striped pattern, changes color or turns darker when exposed to the sterilisation process.