PTQ10, Organic Photovoltaics (OPV) reagent

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Poly[(thiophene)-alt-(6,7-difluoro-2-(2-hexyldecyloxy)quinoxaline)]

H66106.MC H66106.MD H66106.ME
H66106.MCEA 504 EUR
H66106.MC H66106.MD H66106.ME
PTQ10, Organic Photovoltaics (OPV) reagent
PTQ10
Formule: (C₂₈H₃₆F₂N₂OS)n
CAS nummer: 2270233-86-6

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1,96 ± 0,02 eV


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR