Tuerk solution for microscopy, for leukocyte count, Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Turk solution, Acetic acid gentian violet solution
M-Clarity™ quality level = MQ100
93770-100ML 93770-1L
93770-100MLEA 28.7 EUR
93770-100ML 93770-1L
Tuerk solution for microscopy, for leukocyte count, Sigma-Aldrich®
Tuerk solution
Counting leukocytes in the counting chamber.