Water, molecular grade DNAse_RNAse_free
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Hydrogen oxide
786-72C 786-73C 786-292 786-293
786-72CEA 231 EUR
786-72C 786-73C 786-292 786-293
Water, molecular grade DNAse_RNAse_free
Water
Molecular Grade Water is ultrapure water that certified RNase, DNase and Protease free, is suitable for use in molecular biology applications, which demand a high quality of water and assurance that the water is free from nucleases and protease contamination. No toxic agents, such as DEPC, are used in the manufacturing of Molecular Grade Water, eliminating DEPC interference of enzymatic reactions.
  • Each lot is quality tested for the absence of RNase, DNase and protease contamination
For use in molecular biology applications, PCR reactions, 2D electrophoresis and mass spectrometry.
Formule: H₂O
MW: 18.02 g/mol
Kookpunt: 100
Smeltpunt: 0
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011332
CAS nummer: 7732-18-5
Merck Index: 13,10098
Order Now
Specificaties
Environmentally Preferable