Ruthenium(III) chloride oxide 85.0-115.0% dried basis, AT Microscopy, Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Ruthenium oxychloride, Tetraamminechlorohydroxoruthenium(III) chloride, Ruthenium Red
00541-1G 00541-5G
00541-1GEA 284 EUR
00541-1G 00541-5G
Ruthenium(III) chloride oxide 85.0-115.0% dried basis, AT Microscopy, Sigma-Aldrich®
Ruthenium(III) chloride oxide
(Ammoniated ruthenium oxychloride)
Formule: H₄₂Cl₆N₁₄O₂Ru₃
MW: 786.354 g/mol
MDL Nummer: MFCD00011479
CAS nummer: 11103-72-3