Control slide, Fungi TISSUE-TROL™, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TISSUE-TROL™
TTR004-25EAEA 248 EUR
TTR004-25EA
Control slide, Fungi TISSUE-TROL™, Sigma-Aldrich®
Object- en dekglaasjes Microscopy Control Slides
Histology slides with paraffin embedded section of mouse lung tissue containing Candida albicans.
Suitable for use with Silver Stain (Modified GMS) Procedure No. HT100.