Control slide, Melanin TISSUE-TROL™, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TISSUE-TROL™
TTR014-25EAEA 288 EUR
TTR014-25EA
Control slide, Melanin TISSUE-TROL™, Sigma-Aldrich®
Object- en dekglaasjes Microscopy Control Slides
Histology slides with paraffin embedded section of human skin containing melanin.
Suitable for use with Fontana-Masson Stain Kit (Procedure No. HT200).