Cone folded filter papers, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
990010112 990010116 990110116 1002-10240
WHAT990010112EA 829 EUR
WHAT990010112 WHAT990010116 WHAT990110116 WHAT1002-10240
Cone folded filter papers, Whatman™
Filters Filterpapier Kwalitatief filterpapier
Cone folded filter paper saves valuable time when undertaking repetitive or multiple analyses.
  • Easily fits into conical funnels
  • Saves valuable time when undertaking repetitive or multiple analyses
  • Provides the same high quality and consistency that you expect from all Whatman filter papers
Whatman cellulose filter paper grades are available in ready-to-use cone folded formats.
Order Now
Specificaties
Nr 40 1 2
Environmentally Preferable
Dikte 210 µm 180 µm
Asgehalte 0,007% 0,06%
Gewicht 95 g/m² 87 g/m²