Molecular detection instrument and accessory kit
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MMMPMDS100EUEA 23660 EUR
MMMPMDS100EU
Molecular detection instrument and accessory kit
testen Immunologie-assays
The 3M™ Molecular detection instrument comes with a pluggable heater, software, power cord, USB cable and a basic accessory kit.
  • Smaller footprint than a standard notebook computer that requires significantly less space than the market leading rapid methods
  • Robust instrument that is solid state, portable with no need for recalibration, requires minimal maintenance, and provides automatic diagnostics at start up
  • Powerful software that allows operation of up to four instruments simultaneously from one computer
The basic accessory kit includes a chill block insert, chill block tray, speed loader tray, cap/decap tool lysis, cap/decap tool reagent, and empty lysis and reagent trays.