BL21 competent cells
Leverancier: STRATAGENE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
200132 230240 230245 230250 230255 230265 230275 230280 200131 200132 200133 230130 230132 230134 230240 230245 230250 230255 230280 235200
STRA200132EA 0 EUR
STRA200132 STRA230240 STRA230245 STRA230250 STRA230255 STRA230265 STRA230275 STRA230280 AGLS200131 AGLS200132 AGLS200133 AGLS230130 AGLS230132 AGLS230134 AGLS230240 AGLS230245 AGLS230250 AGLS230255 AGLS230280 AGLS235200
BL21 competent cells
Competente cellen
BL21 competent cells are an all-purpose strain for high-level protein expression and easy induction.
  • High-level protein expression and easy induction
  • Can be used with non-T7 RNA polymerase protein expression systems
  • Allows the tightest control of protein expression for extremely toxic proteins
  • Suitable for expressing proteins from all species, including mammals
  • Ideal for difficult protein expression, especially when codon bias is a problem
  • Increases transformation efficiency with Hte phenotype
  • Derived from Agilent’s high performance BL21-Gold competent
More Product Information
Order Now
Specificaties
Environmentally Preferable
Genotypes