XL10-Gold ultracompetent cells
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HEWL200315EA 0 EUR
HEWL200315 STRA200317 STRA200314 STRA200315 AGLS200314 AGLS200315
XL10-Gold ultracompetent cells
Competente cellen
XL10-Gold ultracompetent cells transform large plasmids and ligated DNA with highest efficiency.
  • Engineered to transform large plasmids and ligated DNA more efficiently than other cell lines
  • Includes Hte phenotype for 20-fold to 30-fold higher transformation efficiency of ligated DNA and better competent cell performance
  • Chemically competent cells that allow efficient transformation of large DNA molecules, including expression vectors and genomic DNA
  • Ideal for plasmid library construction by decreasing bias against larger plasmids and producing more colonies for a more representative library
  • Transformation efficiency of ≥5×10⁹ transformants/µg
The XL10-Gold ultracompetent cells were designed for cloning large plasmids and ligated DNA with the highest transformation efficiency possible, while exhibiting faster growth and larger colonies. XL10-Gold Ultracompetent Cells possess the high transformation phenotype (Hte), which gives these cells an average transformation efficiency of ≥5×10⁹ transformants/μg of supercoiled DNA Plasmid libraries constructed in this strain are more representative because XL10-Gold cells decrease the bias against large inserts.
Opgelet: For research use only. Not for use in diagnostic procedures.