Graduated wide neck bottles, PE, with screw cap and undercap
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
408 409 411 412 413 410
216-2992EA 468 EUR
216-2992 216-2993 216-2507 216-2995 216-2996 216-2994
Graduated wide neck bottles, PE, with screw cap and undercap
Flessen Kunststof flessen
These high quality graduated wide neck bottles are made of PE (main body and under cap) and supplied with a PP cap.
  • Suitable for foodstuff contact according to EC Regulation 10/2011
  • Translucent
  • Tagging points on shoulder and cap ensures tamper-evidence by means of tags and seals
Graduated bottle for liquids and powders. Sloping shoulders reduce tendency for residue deposits. Moulded-in graduations.
Certificaten: Meets EC Regulation 10/2011. Migration Test Compliance Declaration available.