SafetyCaps, V2.0, GL 45, preparative
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
307007 307008 307009 308032 309032 310032 307003 309034
SCTV307007EA 129 EUR
SCTV307007 SCTV307008 SCTV307009 SCTV308032 SCTV309032 SCTV310032 SCTV307003 SCTV309034
SafetyCaps, V2.0, GL 45, preparative
Sluitingen Veiligheidsdoppen en veiligheidsafvaldoppen
Special version of the SafetyCaps V2.0 for use in the preparative HPLC environment with larger tubing connections and air valves with a higher flow rate.
  • Standard with GL 45 thread
  • Higher flow rate of the valve with up to 400 ml/min
  • Larger tubing connections from OD 3,2 mm to 6,35 mm
The SafetyCaps V2.0, preparative are equipped with a red air valve and are specially designed for preparative HPLC operation. They allow flow rates of up to 400 ml per minute without any problems, whereas the standard blue vent valves used are only limited to a flow rateMore Product Information
Order Now
Specificaties
Environmentally Preferable