PCR polishing kit
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AGLS200409EA 0 EUR
AGLS200409
PCR polishing kit
Reagentia voor nucleïnezuren Nucleic Acid Modification and Labelling
PCR polishing kit increases blunt-end cloning efficiencies associated with PCR-generated fragments.
  • Generates blunt-ended DNA fragments in 30 minutes
  • More efficient than Klenow polishing
  • Optimizes blunt-end cloning efficiencies
PCR Polishing Kit enables an increase in blunt-ended cloning efficiencies associated with polymerase chain reaction (PCR)-generated fragments. It polishes the ends of the 3’ overhang extensions of polymerase-generated DNA fragments directly from PCR amplification and can completely fill in 5’ overhangs to generate blunt ends.
Dramatic increases in the population of blunt-end DNA fragments following polishing with Agilent’s PCR Polishing Kit enables significant increase in overall experimental efficiency associated with procedures utilising blunt-ended ligation.