PCR and qPCR consumables
Leverancier: STRATAGENE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
410088 410090 410091 410092 410098 401411 410094 401333 401334
STRA410088EA 0 EUR
STRA410088 STRA410090 STRA410091 STRA410092 STRA410098 STRA401411 HEWL410094 AGLS401333 AGLS401334
PCR and qPCR consumables
PCR-toestellen
High-quality well plate supplies and accessories will enhance your PCR and qPCR workflow and analysis.
Agilent AriaMx low profile strip tubes are optimised for PCR and qPCR applications. The tubes are validated for optimal performance with Mx3005P, Mx3000P, and the AriaMx real time PCR Systems. Part of the qPCR Plastics and Accessories, the products include the perfect fit frame, tubes, and caps.
Additional products available include the 96-well semiskirted polypropylene PCR plate compatible with the optical Strip Caps, the StrataCoolers LP Benchtop Cooler, and StrataCooler cryo preservation module. Tubes and accessories are compatible most PCR and Real-Time instruments.
Opgelet: For research use only. Not for use in diagnostic procedures.