VWR® PESTINORM®, Gas Chromatography (GC) Certified Reference Material
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
91000.050EA 215 EUR
91000.050 91003.050 91008.050 91009.050 91010.050 91012.050 91016.050 91017.050 91019.050 91030.050 91032.050 91036.050 91038.050 91039.050 91040.050 91041.050 91042.050 91044.050 91045.050 91046.050 91047.050 91050.050 91053.050 91055.050 91061.050 91062.050 91064.050 91066.050 91067.050 91068.050 91069.050
VWR® PESTINORM®, Gas Chromatography (GC) Certified Reference Material
Standaarden Standaarden voor chromatografie
Due to its excellent separation capacity and its high sensitivity gas chromatography is a widely used technique for analytical purposes. In order to get proper identification and quantification results of the analysed compounds, reference standards are needed. These PESTINORM® reference substances can be used for the characterisation of GC column properties. In addition they may also be used for the identification of unknown compounds in a gas chromatogram or as standards in quantitative GC analysis.
  • All products are CRM acc ISO/IEC 17025 and ISO 17034
  • All products supplied with a Certificate of Analysis and batch-specific chromatogram
  • Assay typically min 99,0% in many cases over 99,5%
  • 24 month shelf life
Intended uses:
- For calibration of TLC, GC-FID, GC-TCD, GC-ECD, GC-MS, GC-MS-MS, LC-UV, LC-MS and LC-MS-MS
- Validation of analytical methods
- Preparation of 'working reference samples'
- Detection limit and linearity studies