4-Nitrophenyl-β-D-lactopyranoside
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIB6128-1G
BIB6128-500MG
APOSBIB6128-1GEA
970
EUR
APOSBIB6128-1G
APOSBIB6128-500MG
4-Nitrophenyl-β-D-lactopyranoside
4-Nitrophenyl-β-D-lactopyranoside
Formule:
C₁₈H₂₅NO₁₃ Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD00069843 CAS nummer: 4419-94-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...