1,6:2,3-Dianhydro-4-O-(2,3-di-O-benzyl-β-D-glucopyranosyl)-β-D-mannopyranose ≥99%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BICL4086-100MG BICL4086-250MG BICL4086-50MG
APOSBICL4086-100MGEA 548 EUR
APOSBICL4086-100MG APOSBICL4086-250MG APOSBICL4086-50MG
1,6:2,3-Dianhydro-4-O-(2,3-di-O-benzyl-β-D-glucopyranosyl)-β-D-mannopyranose ≥99%
1,6:2,3-Dianhydro-4-O-(2,3-di-O-benzyl-β-D-glucopyranosyl)-β-D-mannopyranose