6-O-(Carboxymethyl)-D-glucose ≥95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BICL4289-2MG BICL4289-5MG BICL4289-1MG
APOSBICL4289-2MGEA 380 EUR
APOSBICL4289-2MG APOSBICL4289-5MG APOSBICL4289-1MG
6-O-(Carboxymethyl)-D-glucose ≥95%
6-O-(Carboxymethyl)-D-glucose
Formule: C₈H₁₄O₈
MW: 238.192 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28122842
CAS nummer: 95350-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR