4-Methylphenyl 3-O-benzoyl-4,6-O-benzylidene-1-thio-α-D-mannopyranoside ≥98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BICL4359-100MG BICL4359-250MG BICL4359-500MG
APOSBICL4359-100MGEA 563 EUR
APOSBICL4359-100MG APOSBICL4359-250MG APOSBICL4359-500MG
4-Methylphenyl 3-O-benzoyl-4,6-O-benzylidene-1-thio-α-D-mannopyranoside ≥98%
4-Methylphenyl 3-O-benzoyl-4,6-O-benzylidene-1-thio-α-D-mannopyranoside