Alexidine dihydrochloride
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1,1'-Hexamethylenebis[5-(2-ethylhex-1-yl)]biguanide dihydrochloride
BIMI8260-1G BIMI8260-250MG
APOSBIMI8260-1GEA 198 EUR
APOSBIMI8260-1G APOSBIMI8260-250MG
Alexidine dihydrochloride
Alexidine dihydrochloride
Alexidine dihydrochloride is a selective and potent inhibitor of protein tyrosine phosphatases localized to mitochondrion 1 (PTPMT1). It has antibacterial, antiplaque properties against Streptococcus mutans, Actinomyces ciscosus, and Actinomyces naesludii.
Formule: C₂₆H₅₆N₁₀·2ClH
MW: 581.712 g/mol
Smeltpunt: 219…221 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00082369
CAS nummer: 1715-30-6