L-α-Phosphatidylinositol from Glycine max (soybean)

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1,2-Diacyl-sn-glycero-3-phospho-(1-D-myo-inositol)

BIP0140-1G BIP0140-250MG
APOSBIP0140-1GEA 672 EUR
APOSBIP0140-1G APOSBIP0140-250MG
L-α-Phosphatidylinositol from Glycine max (soybean)
L-α-Phosphatidylinositol

Phospholipid membrane component, precursor of inositol phosphates

Formule: C₄₅H₈₇O₁₃P
MW: 867.138 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00151139
CAS nummer: 97281-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR