5-Bromo-1,4,6-trimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR110358-1G OR110358-250MG
APOSOR110358-1GEA 141 EUR
APOSOR110358-1G APOSOR110358-250MG
5-Bromo-1,4,6-trimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
5-Bromo-1,4,6-trimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Formule: C₉H₉BrN₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16619587
CAS nummer: 1379301-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR