3-[2-(4-tert-Butoxycarbonylpiperazin-1-yl)ethoxy]acetophenone
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR110566-1G
OR110566-10G
OR110566-5G
APOSOR110566-1GEA
199
EUR
APOSOR110566-1G
APOSOR110566-10G
APOSOR110566-5G
3-[2-(4-tert-Butoxycarbonylpiperazin-1-yl)ethoxy]acetophenone
3-[2-(4-tert-Butoxycarbonylpiperazin-1-yl)ethoxy]acetophenone
Formule:
C₁₉H₂₈N₂O₄ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD11109538 CAS nummer: 351076-06-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...