tert-Butyl 4-(3-methoxy-3-oxopropyl)homopiperazine-1-carboxylate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR110608-1G OR110608-10G OR110608-5G
APOSOR110608-1GEA 249 EUR
APOSOR110608-1G APOSOR110608-10G APOSOR110608-5G
tert-Butyl 4-(3-methoxy-3-oxopropyl)homopiperazine-1-carboxylate
tert-Butyl 4-(3-methoxy-3-oxopropyl)homopiperazine-1-carboxylate
Formule: C₁₄H₂₆N₂O₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD05664953
CAS nummer: 874801-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR