Ethyl 5-[(acetyloxy)methyl]-4-ethyl-3-methyl-1H-pyrrole-2-carboxylate
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR110639-1G
OR110639-10G
APOSOR110639-1GEA
199
EUR
APOSOR110639-1G
APOSOR110639-10G
Ethyl 5-[(acetyloxy)methyl]-4-ethyl-3-methyl-1H-pyrrole-2-carboxylate
Ethyl 5-[(acetyloxy)methyl]-4-ethyl-3-methyl-1H-pyrrole-2-carboxylate
Formule:
C₁₃H₁₉NO₄ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD01240570 CAS nummer: 100619-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...