3-Bromo-2-methylimidazo[1,2-a]pyridine
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR111096-1G OR111096-5G OR111096-250MG
APOSOR111096-1GEA 87.6 EUR
APOSOR111096-1G APOSOR111096-5G APOSOR111096-250MG
3-Bromo-2-methylimidazo[1,2-a]pyridine
3-Bromo-2-methylimidazo[1,2-a]pyridine
Formule: C₈H₇BrN₂
MDL Nummer: MFCD02251329
CAS nummer: 4805-70-3