3-Chloro-N,N-diisopropylthiophene-2-carboxamide
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR111136-1G
OR111136-5G
APOSOR111136-1GEA
274
EUR
APOSOR111136-1G
APOSOR111136-5G
3-Chloro-N,N-diisopropylthiophene-2-carboxamide
3-Chloro-N,N-diisopropylthiophene-2-carboxamide
Formule:
C₁₁H₁₆ClNOS Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD28987628 CAS nummer: 1980049-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...