N,N-Dimethyl-1,3,4-thiadiazole-2,5-diamine
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR111194-10G OR111194-1G OR111194-5G
APOSOR111194-10GEA 0 EUR
APOSOR111194-10G APOSOR111194-1G APOSOR111194-5G
N,N-Dimethyl-1,3,4-thiadiazole-2,5-diamine
N,N-Dimethyl-1,3,4-thiadiazole-2,5-diamine
Formule: C₄H₈N₄S
MDL Nummer: MFCD18805198
CAS nummer: 72269-96-6