8-Chloronaphthalene-1-sulphonic acid 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
OR21393-1G
OR21393-250MG
OR21393-5G
APOSOR21393-1GEA
58.4
EUR
APOSOR21393-1G
APOSOR21393-250MG
APOSOR21393-5G
8-Chloronaphthalene-1-sulphonic acid 97%
8-Chloronaphthalene-1-sulphonic acid
Formule:
C₁₀H₇ClO₃S MW: 242.679 g/mol Smeltpunt: 145…147 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00052379 CAS nummer: 145-74-4 EINECS: 205-661-0 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...