Thiophene-3,4-dicarbonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR2655-1G OR2655-5G
APOSOR2655-1GEA 24.6 EUR
APOSOR2655-1G APOSOR2655-5G
Thiophene-3,4-dicarbonitrile
Thiophene-3,4-dicarbonitrile
Formule: C₆H₂N₂S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD01233047
CAS nummer: 18853-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR